Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals
Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043245.36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS57219-64-4
IUPAC Name[({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
Molecular FormulaCH2O7Zr2
InChI KeyACWOHNQUPJAZIT-UHFFFAOYSA-J
SMILESO[Zr](=O)OC(=O)O[Zr](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)37% ZrO₂ min
Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only