Thermo Scientific Chemicals

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

Catalog number: 043245.A3
2 kg, Each
Thermo Scientific Chemicals

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

Catalog number: 043245.A3
2 kg, Each
Quantity
Catalog number: 043245.A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
57219-64-4
IUPAC Name
[({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
Molecular Formula
CH2O7Zr2
InChI Key
ACWOHNQUPJAZIT-UHFFFAOYSA-J
SMILES
O[Zr](=O)OC(=O)O[Zr](O)=O
Assay (unspecified)
37% ZrO₂ min

Description

Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text