Thermo Scientific Chemicals

n-Hexane, anhydrous, Thermo Scientific Chemicals

Catalog number: 043263.K2
1 L, Each
Thermo Scientific Chemicals

n-Hexane, anhydrous, Thermo Scientific Chemicals

Catalog number: 043263.K2
1 L, Each
Quantity
Catalog number: 043263.K2
also known as 043263-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-54-3
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Residue on Evaporation
≤5ppm
Impurity content
Sulfur compounds (as Sulfur) ≤0.005%
Water Content (Karl Fischer Titration)
≤0.02%
Assay (GC)
≥99.5% (as C6 Hydrocarbons)
Impurity content
Thiophene (Passes test)

Description

n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text