Thermo Scientific Chemicals

Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 043282.K4
2.5 L, Each
Thermo Scientific Chemicals

Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 043282.K4
2.5 L, Each
Quantity
Catalog number: 043282.K4
also known as 043282-K4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-50-3
IUPAC Name
trimethylamine
Molecular Formula
C3H9N
InChI Key
GETQZCLCWQTVFV-UHFFFAOYSA-N
SMILES
CN(C)C
Appearance (Color)
Clear colorless
Assay (Titration)
43-49%
Form
Liquid
Refractive Index
1.3660-1.3720 @ 20?C

Description

Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text