Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals

Catalog number: 043292.14
25 g, Each
Thermo Scientific Chemicals

Poly(styrene-divinylbenzene), 1% cross-linked, 100-200 mesh, Thermo Scientific Chemicals

Catalog number: 043292.14
25 g, Each
Quantity
Catalog number: 043292.14
also known as 043292-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-70-7
MDL Number
9003-70-7
Appearance (Color)
White
Form
Beads
Particle Size
100-200 Mesh
Assay (special)
DVB: 1%

Description

Poly(styrene-divinylbenzene) is used in chromatographic separation medium. It is also used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether and tert-butylcatechol. It is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions. Styrene-divinylbenzene copolymer is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text