Molybdenum wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Molybdenum wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 043311.G1
also known as 043311-G1
Price (USD)/ Each
63.30
-
Add to cart
Quantity:
1 m
Request bulk or custom format
Price (USD)/ Each
63.30
Add to cart
Molybdenum wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number043311.G1
Price (USD)/ Each
63.30
-
Add to cart
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.6mm (0.063in) diameter, annealed.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only