Tungsten wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Tungsten wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number 043312.G5
also known as 043312-G5
Price (USD)/ Each
396.65
Online exclusive
441.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
396.65
Online exclusive
441.00 
Save 44.35 (10%)
Add to cart
Tungsten wire, 1.6mm (0.063in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number043312.G5
Price (USD)/ Each
396.65
Online exclusive
441.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
FormWire, 1.6mm (0.063in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only