Thermo Scientific Chemicals

9,10-Bis(phenylethynyl)anthracene, Thermo Scientific Chemicals

Catalog number: 043313.03
1 g, Each
Thermo Scientific Chemicals

9,10-Bis(phenylethynyl)anthracene, Thermo Scientific Chemicals

Catalog number: 043313.03
1 g, Each
Quantity
Catalog number: 043313.03
also known as 043313-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10075-85-1
IUPAC Name
9,10-bis(2-phenylethynyl)anthracene
Molecular Formula
C30H18
InChI Key
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C#CC1=C2C=CC=CC2=C(C#CC2=CC=CC=C2)C2=CC=CC=C12
Appearance (Color)
Yellow to orange
Assay (HPLC)
> 97.0%
Form
Powder

Description

9,10-Bis(phenylethynyl)anthracene is used as a chemiluminescent fluorophore with high quantum efficiency. It is used in lightsticks as a fluorophor producing ghostly green light. It is also used as a dopant for organic semiconductors in OLEDs. It is used as a reagent for chemiluminescence research. 9,10-Bis(phenylethynyl)anthracene is known to exert fluorescence properties due to its polycyclic aromatic hydrocarbon system and it is commonly used as an scintillator additive. 9,10-Bis(phenylethynyl)anthracene was also studied extensively as a fluorescence emitters for peroxyoxalate chemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text