Thermo Scientific Chemicals
2-Butanol, anhydrous, 99%, Thermo Scientific Chemicals
Catalog number: 043315.K2
1 L, Each
Thermo Scientific Chemicals
2-Butanol, anhydrous, 99%, Thermo Scientific Chemicals
Catalog number: 043315.K2
1 L, Each
Quantity
Catalog number: 043315.K2
also known as 043315-K2
Price (USD)
Quantity
-
Have Questions?
Description
2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.