Thermo Scientific Chemicals

Brilliant Blue G, ultrapure

Catalog number: 043318.04
2 g, Each
Thermo Scientific Chemicals

Brilliant Blue G, ultrapure

Catalog number: 043318.04
2 g, Each
Quantity
Catalog number: 043318.04
also known as 043318-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6104-58-1
IUPAC Name
sodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}-2-methylphenyl)methylium
Molecular Formula
C47H48N3NaO7S2
InChI Key
CMGWKYXJIYYESN-UHFFFAOYSA-M
SMILES
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2C)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
Form
Powder
Comment
C.I. 42655
Loss on Drying
≤10% (105?C, 3h)
Assay (Dye)
≥65%
Comment
Suitable for protein staining

Description

Brilliant Blue G is used in analytical biochemistry dye and as a P2X7 antagonist. It is also used in protein staining following gel electrophoresis. It is used to prepare the protein reagent for the determination of protein content of the collagenase enzyme isolated from fish waste. It may be employed as stain for the internal limiting membrane (ILM) for the macular hole (MH) and epiretinal membrane (ERM) surgery. It is useful for SDS gels as it readily stains proteins with minimal background color. Protein bands can be visualized during staining. It has been used in the Bradford dye-binding protein assay. A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids. Brilliant Blue G has been used for determining critical micelle concentration of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text