Thermo Scientific Chemicals

Tetra-n-butylammonium nitrate, 98%, Thermo Scientific Chemicals

Catalog number: 043342.22
100 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium nitrate, 98%, Thermo Scientific Chemicals

Catalog number: 043342.22
100 g, Each
Quantity
Catalog number: 043342.22
also known as 043342-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1941-27-1
IUPAC Name
tetrabutylazanium nitrate
Molecular Formula
C16H36N2O3
InChI Key
QHOKENWFMZXSEU-UHFFFAOYSA-N
SMILES
[O-][N+]([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
Off-white to faintly yellow
Assay (Aqueous acid-base Titration)
> 97.0%
Form
powder
Water Content (Karl Fischer Titration)
1.0% max

Description

Tetra-n-butylammonium nitrate is used in coatings, printing ink, rubber, glass, leather and cosmetics. It serves as a supporting electrolyte in liquid ammonia. It is also used in the preparation of nitrates from triflates. It is employed in the inversion of configuration of alcohols through tosylates. It is also a source of NO3- used in nucleophilic substitution reactions. Further, it is used in the conversion of halides into ketones. In addition to this, it is used as a source of nitronium ions for the nitration of olefins, amides, ribonucleosides, glycols and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text