Thermo Scientific Chemicals
3-Hydroxybutyric acid, tech., Thermo Scientific Chemicals
Catalog number: 043354.06
5 g, Each
Thermo Scientific Chemicals
3-Hydroxybutyric acid, tech., Thermo Scientific Chemicals
Catalog number: 043354.06
5 g, Each
Quantity
Catalog number: 043354.06
also known as 043354-06
Price (USD)
Quantity
-
Have Questions?
Description
Industrially, 3-hydroxybutanoic acid is used for the manufacture of biodegradable plastics. It is also acts as a solvent in organoleptics, cosmetics. It is also used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by Escherichia coli.