Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals
Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Catalog number 043359.K4
also known as 043359-K4
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals
Catalog number043359.K4
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS79-10-7
IUPAC Nameprop-2-enoic acid
Molecular FormulaC3H4O2
InChI KeyNIXOWILDQLNWCW-UHFFFAOYSA-N
SMILESOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay from Suppliers CofA≥99.4% (Assay: GC)
CommentMay contain various amounts of Diacrylic acid (Assay: LC)
Water Content (Karl Fischer Titration)≤0.1%
View more
Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only