Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min, Thermo Scientific Chemicals
Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 043360.06
also known as 043360-06
Price (USD)/ Each
790.65
Online exclusive
878.00 
Save 87.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
790.65
Online exclusive
878.00 
Save 87.35 (10%)
Add to cart
Gold(III) chloride, Premion™, 99.99% (metals basis), Au 64.4% min, Thermo Scientific Chemicals
Catalog number043360.06
Price (USD)/ Each
790.65
Online exclusive
878.00 
Save 87.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13453-07-1
IUPAC Namegold hydrochloridyl
Molecular FormulaAuCl
InChI KeyBRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES[Cl].[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Au: ≥ 64.4%
Appearance (Color)Brown-red
FormCrystalline
Total Metal Impurities≤0.01%
Gold(III) chloride is used in colloidal gold solutions, in photography and as a print toning agent(gold toning), starting solution to form other gold compounds and a precursor for preparation of ultra pure gold metal. It is used in electroplating and electroless plating as an anode in an electric cell. Gold(III) chloride acts as a gold catalyst and cell body stains for bright field and dark field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only