Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis), Thermo Scientific Chemicals
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Pc.
Catalog number 043361.KG
Price (USD)/ Each
529.00
-
Add to cart
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)/ Each
529.00
Add to cart
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043361.KG
Price (USD)/ Each
529.00
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormPlate, 3.175mm (0.125in) thick.
Assay from Supplier's CofA≥99.99% (metals basis)
Width100mm (3.9in) width.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only