Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 043364.04
also known as 043364-04
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
Add to cart
Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043364.04
Price (USD)/ Each
153.65
Online exclusive
170.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14898-67-0
IUPAC Nameruthenium(3+) trichloride
Molecular FormulaCl3Ru
InChI KeyYBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ru+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Ruthenium(III) chloride hydrate is used as a starting material in ruthenium chemistry. It acts as a precursor to many ruthenium compounds, which finds application as catalysts in asymmetric hydrogenation and alkene metathesis. It serves as a catalyst in the preparation of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine. Further, it is used in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst for the efficient dehydrogenation of arylmethyl alcohols to the corresponding aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only