Thermo Scientific Chemicals

Gold(I) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043365.06
5 g, Each
Thermo Scientific Chemicals

Gold(I) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043365.06
5 g, Each
Quantity
Catalog number: 043365.06
also known as 043365-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-29-8
IUPAC Name
gold(1+) chloride
Molecular Formula
AuCl
InChI Key
FDWREHZXQUYJFJ-UHFFFAOYSA-M
SMILES
[Cl-].[Au+]
Total Metal Impurities
≤0.1%
Appearance (Color)
Yellow
Assay (special)
Au 84.4-84.7%
Form
Powder

Description

Gold(I) chloride is used as a starting material for the preparation of Phosphine ligated gold(I) complexes, which is used as catalysts in synthetic chemistry. It is also used to prepare metallic gold and potassium auric bromide and potassium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text