Thermo Scientific Chemicals

Lithium tungsten oxide, 99% min, Thermo Scientific Chemicals

Catalog number: 043366.18
50 g, Each
Thermo Scientific Chemicals

Lithium tungsten oxide, 99% min, Thermo Scientific Chemicals

Catalog number: 043366.18
50 g, Each
Quantity
Catalog number: 043366.18
also known as 043366-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13568-45-1
IUPAC Name
dilithium(1+) dioxotungstenbis(olate)
Molecular Formula
Li2O4W
InChI Key
SCKKBXOJPMZVPF-UHFFFAOYSA-N
SMILES
[Li+].[Li+].[O-][W]([O-])(=O)=O
Assay (unspecified)
99% min

Description

Lithium tungsten oxide is employed for the preparation of ceramics with ultra-low sintering temperatures. It acts as a catalyst for oxidative coupling reactions. Furthermore, it is used in the production of high density aqueous polytungstate (metatungstate) solutions which are utilized for the separation of minerals and other solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text