Thermo Scientific Chemicals

Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043374.06
5 g, Each
Thermo Scientific Chemicals

Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043374.06
5 g, Each
Quantity
Catalog number: 043374.06
also known as 043374-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10476-86-5
IUPAC Name
strontium(2+) diiodide
Molecular Formula
I2Sr
InChI Key
KRIJWFBRWPCESA-UHFFFAOYSA-L
SMILES
[Sr++].[I-].[I-]
Total Metal Impurities
0.01% max.

Description

Strontium iodide is used as a scintillation gamma radiation detector. It is also used in a range of hand-held radiation detection instruments as well as in medical, industrial and environmental applications. It is used in medicine as a substitute for potassium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text