Thermo Scientific Chemicals

Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 043377.14
25 g, Each
Thermo Scientific Chemicals

Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog number: 043377.14
25 g, Each
Quantity
Catalog number: 043377.14
also known as 043377-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-34-7
IUPAC Name
oxo[(oxovanadio)oxy]vanadium
Molecular Formula
O3V2
InChI Key
KFAFTZQGYMGWLU-UHFFFAOYSA-N
SMILES
O=[V]O[V]=O
Total Metal Impurities
0.3% max.

Description

Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text