Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 043377.36
also known as 043377-36
Price (USD)/ Each
1,159.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,159.00
Add to cart
Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Catalog number043377.36
Price (USD)/ Each
1,159.00
-
Add to cart
Chemical Identifiers
CAS1314-34-7
IUPAC Nameoxo[(oxovanadio)oxy]vanadium
Molecular FormulaO3V2
InChI KeyKFAFTZQGYMGWLU-UHFFFAOYSA-N
SMILESO=[V]O[V]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only