Thermo Scientific Chemicals

Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min, Thermo Scientific Chemicals

Catalog number: 043380.06
5 g, Each
Thermo Scientific Chemicals

Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min, Thermo Scientific Chemicals

Catalog number: 043380.06
5 g, Each
Quantity
Catalog number: 043380.06
also known as 043380-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-83-9
IUPAC Name
iridium(3+) trichloride
Molecular Formula
Cl3Ir
InChI Key
DANYXEHCMQHDNX-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ir+3]
Total Metal Impurities
0.01% max.
Assay (unspecified)
Ir 63.9% min

Description

Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text