Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm, Thermo Scientific Chemicals
Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen &lt;1000ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number 043385.36
also known as 043385-36
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
Add to cart
Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm, Thermo Scientific Chemicals
Catalog number043385.36
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1%
Particle SizeReport Mesh Size Results
FormPowder, -170 to +400 mesh
Impurity contentOxygen: ≤1000 ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only