Thermo Scientific Chemicals

Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 043393.09
10 g, Each
Thermo Scientific Chemicals

Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 043393.09
10 g, Each
Quantity
Catalog number: 043393.09
also known as 043393-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34622-08-7
IUPAC Name
neodymium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9NdO9S3
InChI Key
WYRSPTDNOIZOGA-UHFFFAOYSA-K
SMILES
[Nd+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Assay (unspecified)
97.6% min

Description

Neodymium(III) trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes and as a catalyst for dehydration of fructose, Diels-Alder reactions of cyclopentadiene and alkyl acrylates, α-amination reactions of pyrazolones, polymerization reactions of cyclosiloxanes, direct polycondensation of dicarboxylic acids and diols, regioselective reductive ring opening of benzylidene acetals, Paal-Knorr cyclocondensation and nitration of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text