Thermo Scientific Chemicals

Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals

Catalog number: 043396.03
1 g, Each
Thermo Scientific Chemicals

Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals

Catalog number: 043396.03
1 g, Each
Quantity
Catalog number: 043396.03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12030-49-8
IUPAC Name
iridium tetrahydroxide
Molecular Formula
H4IrO4
InChI Key
UOYXEZMXOVJLIH-UHFFFAOYSA-J
SMILES
[OH-].[OH-].[OH-].[OH-].[Ir]
Total Metal Impurities
≤0.01%
Form
Powder
Appearance (Color)
Black
Assay (special)
Ir 84.5-85.7%

Description

Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text