Potassium chloride, 3M aq. soln., Thermo Scientific Chemicals
Potassium chloride, 3M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium chloride, 3M aq. soln., Thermo Scientific Chemicals

For Refilling Calomel Electrodes
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 043398.AK
also known as 043398-AK
Price (USD)/ Each
30.65
Online exclusive
34.40 
Save 3.75 (11%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
34.40 
Save 3.75 (11%)
Add to cart
Potassium chloride, 3M aq. soln., Thermo Scientific Chemicals
Catalog number043398.AK
Price (USD)/ Each
30.65
Online exclusive
34.40 
Save 3.75 (11%)
-
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA3M
Appearance (Color)Colorless
FormLiquid
For refilling calomel electrodesPotassium chloride, 3M aq. solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only