Thermo Scientific Chemicals

Nickel Iron Copper Molybdenum foil, 0.125mm (0.005 in.) thick, annealed (Mumetal™), Thermo Scientific Chemicals

Catalog number: 043399.FL
75 x 75 mm, Each
Thermo Scientific Chemicals

Nickel Iron Copper Molybdenum foil, 0.125mm (0.005 in.) thick, annealed (Mumetal™), Thermo Scientific Chemicals

Catalog number: 043399.FL
75 x 75 mm, Each
Quantity
Catalog number: 043399.FL
also known as 043399-FL
Price (USD)
Quantity
-
Have Questions?
Appearance (Color)
Silver gray
Assay from Supplier's CofA
Ni:Fe:Cu:Mo; 77:14:5:4 wt%
Form
Foil, 0.125mm (0.005in) thick, annealed.

Description

Nickel Iron Copper Molybdenum foil is used as a magnetic shielding alloy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text