Thermo Scientific Chemicals

Hydroxypropyl cellulose, M.W. 100,000, Thermo Scientific Chemicals

Catalog number: 043400.36
500 g, Each
Thermo Scientific Chemicals

Hydroxypropyl cellulose, M.W. 100,000, Thermo Scientific Chemicals

Catalog number: 043400.36
500 g, Each
Quantity
Catalog number: 043400.36
also known as 043400-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-64-2
Form
Powder
Viscosity
75 to 150 cps ( 5% solution, 25 deg C)
Assay (GC)
53.4 to 80.5% Hydroxyproppoxy group
Appearance (Color)
White to off-white

Description

Thickener, binder, and emulsifier.Hydroxypropyl cellulose is used as an emulsifier, stabilizer, whipping aid and thickener. It is also used as a binder in ceramics and glazes, additive in hair and cosmetic products and blow-molded bottles. Further, it is used in contact lens wetting solutions, as a formulation aid and texturizer for foods. In addition, it serves as a topical ophthalmic protectant and lubricant. It acts as a disintegrant and a binder in tablets. It is also utilized as a sieving matrix for DNA separations by capillary and microchip electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text