Thermo Scientific Chemicals

4-Bromomethyl-6,7-dimethoxycoumarin, 95%, Thermo Scientific Chemicals

Catalog number: 043402.77
0.1 g, Each
Thermo Scientific Chemicals

4-Bromomethyl-6,7-dimethoxycoumarin, 95%, Thermo Scientific Chemicals

Catalog number: 043402.77
0.1 g, Each
Quantity
Catalog number: 043402.77
also known as 043402-77
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88404-25-5
IUPAC Name
4-(bromomethyl)-6,7-dimethoxy-2H-chromen-2-one
Molecular Formula
C12H11BrO4
InChI Key
JGODLBJJCNQFII-UHFFFAOYSA-N
SMILES
COC1=C(OC)C=C2C(CBr)=CC(=O)OC2=C1
Form
powder
Assay (HPLC)
> 94.0%
Appearance (Color)
Yellow

Description

4-Bromomethyl-6,7-dimethoxycoumarin is used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It is also used as derivatization reagent: for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography, in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry and for determination of carboxylic acids in chromatographic detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text