Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals
Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals

Precursor for TaN
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043419.14
also known as 043419-14
Price (USD)/ Each
797.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
938.00 
Save 140.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
797.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
938.00 
Save 140.35 (15%)
Add to cart
Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043419.14
Price (USD)/ Each
797.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
938.00 
Save 140.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialTantalum tris (diethylamido)-tert-butylimide
CAS169896-41-7
Health Hazard 1H261-H314-H335
Health Hazard 2GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
View more
Tantalum tris(diethylamido)-tert-butylimide is used as a precursor for tantalum nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only