Thermo Scientific Chemicals

1-Propanethiol, 98%, Thermo Scientific Chemicals

Catalog number: 043420.AP
500 mL, Each
Thermo Scientific Chemicals

1-Propanethiol, 98%, Thermo Scientific Chemicals

Catalog number: 043420.AP
500 mL, Each
Quantity
Catalog number: 043420.AP
also known as 043420-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-03-9
IUPAC Name
propane-1-thiol
Molecular Formula
C3H8S
InChI Key
SUVIGLJNEAMWEG-UHFFFAOYSA-N
SMILES
CCCS
Appearance (Color)
Colorless
Assay (GC)
≥97.5%
Form
Liquid
Retest Date
1.4350-1.4390 @ 20?C

Description

1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text