1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butanethiol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number 043421.AP
also known as 043421-AP
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
1-Butanethiol, 98%, Thermo Scientific Chemicals
Catalog number043421.AP
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS109-79-5
IUPAC Namebutane-1-thiol
Molecular FormulaC4H10S
InChI KeyWQAQPCDUOCURKW-UHFFFAOYSA-N
SMILESCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay from Supplier's CofA≥97.5%
FormLiquid
1-Butanethiol is used as a model gaseous analyte for fast-responding membrane-free amperometric gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only