Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 500 mm
50 x 50 mm
100 x 100 mm
Catalog number 043424.HP
also known as 043424-HP
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
-
Add to cart
Quantity:
100 x 500 mm
Request bulk or custom format
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
Add to cart
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number043424.HP
Price (USD)/ Each
519.65
Online exclusive
577.00 
Save 57.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metalsless than 0.0005% trace metal impurities
Appearance (Color)Silver
TemperAnnealed
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only