Thermo Scientific Chemicals

Tin(II) bromide, 99.2%, Thermo Scientific Chemicals

Catalog number: 043430.06
5 g, Each
Thermo Scientific Chemicals

Tin(II) bromide, 99.2%, Thermo Scientific Chemicals

Catalog number: 043430.06
5 g, Each
Quantity
Catalog number: 043430.06
also known as 043430-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-24-0
IUPAC Name
λ²-tin(2+) dibromide
Molecular Formula
Br2Sn
InChI Key
ZSUXOVNWDZTCFN-UHFFFAOYSA-L
SMILES
[Br-].[Br-].[Sn++]
Assay (unspecified)
99.2% min

Description

Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text