Ruthenium(III) nitrosylacetate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Ruthenium(III) nitrosylacetate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium(III) nitrosylacetate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 043435.06
also known as 043435-06
Price (USD)/ Each
517.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
517.00
Add to cart
Ruthenium(III) nitrosylacetate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043435.06
Price (USD)/ Each
517.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialRuthenium (III) nitrosylacetate
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
Ruthenium(III) nitrosyl acetate is used to fabricate oxide thin-film transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only