Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 043441.14
also known as 043441-14
Price (USD)/ Each
553.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
553.00
Add to cart
Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043441.14
Price (USD)/ Each
553.00
-
Add to cart
Chemical Identifiers
CAS7718-54-9
IUPAC Namenickel(2+) dichloride
Molecular FormulaCl2Ni
InChI KeyQMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Nickel(II) chloride is used in electroplating, electroforming ,electroless plating, tea and coffee plantation, dye mordant. It is also used in semiconducting metal-containing polymer synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only