Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals

In semiconductor technology
Have Questions?
Quantity:
5 g
Catalog number 043455.06
also known as 043455-06
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
Aluminum antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043455.06
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS25152-52-7
IUPAC Namealumanylidynestibane
Molecular FormulaAlSb
InChI KeyLVQULNGDVIKLPK-UHFFFAOYSA-N
SMILES[Al]#[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
In semiconductor technology

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only