Thermo Scientific Chemicals

Diethylene glycol diethyl ether, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 043464.AK
250 mL, Each
Thermo Scientific Chemicals

Diethylene glycol diethyl ether, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 043464.AK
250 mL, Each
Quantity
Catalog number: 043464.AK
also known as 043464-AK
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-36-7
IUPAC Name
1-ethoxy-2-(2-ethoxyethoxy)ethane
Molecular Formula
C8H18O3
InChI Key
RRQYJINTUHWNHW-UHFFFAOYSA-N
SMILES
CCOCCOCCOCC
Form
Liquid
Specific Absorbance (E 1%/1cm)
260nm ≤ 1.0
Specific Absorbance (E 1%/1cm)
320nm ≤ 0.04
Specific Absorbance (E 1%/1cm)
400nm ≤ 0.01
Appearance (Color)
Colorless

Description

Diethylene glycol diethyl ether is used to study the activities of enzymes in aqueous organic mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text