6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%, Thermo Scientific Chemicals
6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 043467.06
also known as 043467-06
Price (USD)/ Each
306.65
Online exclusive
340.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
306.65
Online exclusive
340.00 
Save 33.35 (10%)
Add to cart
6-Methyl-4-trifluoromethyl-2(1H)-pyridone, 97%, Thermo Scientific Chemicals
Catalog number043467.06
Price (USD)/ Each
306.65
Online exclusive
340.00 
Save 33.35 (10%)
-
Add to cart
Chemical Identifiers
CAS22123-19-9
IUPAC Name6-methyl-4-(trifluoromethyl)-1,2-dihydropyridin-2-one
Molecular FormulaC7H6F3NO
InChI KeyCDRMKCNOGIBXOG-UHFFFAOYSA-N
SMILESCC1=CC(=CC(=O)N1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder or crystals or crystalline powder
Melting Point119.0-122.0°C
Purity≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only