Thermo Scientific Chemicals

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043472.14
25 g, Each
Thermo Scientific Chemicals

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043472.14
25 g, Each
Quantity
Catalog number: 043472.14
also known as 043472-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-46-0
IUPAC Name
λ²-iron(2+) dibromide
Molecular Formula
Br2Fe
InChI Key
GYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES
[Fe++].[Br-].[Br-]
Appearance (Color)
Yellow
Total Metal Impurities
0.005% max.
Residual water
< 100 ppm
Form
Powder

Description

It is used as a Polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text