Thermo Scientific Chemicals

Barium carbonate, tech., Thermo Scientific Chemicals

Catalog number: 043478.22
100 g, Each
Thermo Scientific Chemicals

Barium carbonate, tech., Thermo Scientific Chemicals

Catalog number: 043478.22
100 g, Each
Quantity
Catalog number: 043478.22
also known as 043478-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
513-77-9
IUPAC Name
barium(2+) carbonate
Molecular Formula
CBaO3
InChI Key
AYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES
[Ba++].[O-]C([O-])=O
Assay (unspecified)
95% min

Description

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text