Barium carbonate, tech., Thermo Scientific Chemicals
Barium carbonate, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium carbonate, tech., Thermo Scientific Chemicals

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
10 kg
Catalog number 043478.A3
also known as 043478-A3
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
Barium carbonate, tech., Thermo Scientific Chemicals
Catalog number043478.A3
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95% min
Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only