Graphite flake, median 7-10 micron, 99% (metals basis), Thermo Scientific Chemicals
Graphite flake, median 7-10 micron, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite flake, median 7-10 micron, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043480.A1
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofACarbon, C (by Loss on Ignition): ≥99.0%
Residual water≤0.5%
Surface Area5-15m²/g
Particle Size≥99.5% is -325 mesh (≤44µm)
CommentManufactured from natural high purity graphite flake
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only