Thermo Scientific Chemicals

Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals

Catalog number: 043484.30
250 g, Each
Thermo Scientific Chemicals

Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals

Catalog number: 043484.30
250 g, Each
Quantity
Catalog number: 043484.30
also known as 043484-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13494-90-1
IUPAC Name
gallium(3+) trinitrate
Molecular Formula
GaN3O9
InChI Key
CHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Concentration (weight/weight)
Ga 9-10%

Description

Gallium(III) nitrate solution, is used as the material for producing gallium salts and for scientific research and chemical reagent. It is also used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text