Thermo Scientific Chemicals

Iron phosphide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 043486.09
10 g, Each
Thermo Scientific Chemicals

Iron phosphide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 043486.09
10 g, Each
Quantity
Catalog number: 043486.09
also known as 043486-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12023-53-9
IUPAC Name
triferriophosphane
Molecular Formula
Fe3P
InChI Key
KFSKMQQNHSLZRJ-UHFFFAOYSA-N
SMILES
[Fe]P([Fe])[Fe]
Total Metal Impurities
0.5% max.

Description

Iron phosphide is used as a semiconductor. It finds application in high power and high frequency applications like laser diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text