Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043488.14
also known as 043488-14
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.70 
Save 5.05 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.70 
Save 5.05 (15%)
Add to cart
Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number043488.14
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
32.70 
Save 5.05 (15%)
-
Add to cart
Chemical Identifiers
CAS1327-53-3
IUPAC Namediarsorosooxidane
Molecular FormulaAs2O3
InChI KeyIKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILESO=[As]O[As]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only