Thermo Scientific Chemicals

Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 043488.30
250 g, Each
Thermo Scientific Chemicals

Arsenic(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 043488.30
250 g, Each
Quantity
Catalog number: 043488.30
also known as 043488-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1327-53-3
IUPAC Name
diarsorosooxidane
Molecular Formula
As2O3
InChI Key
IKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILES
O=[As]O[As]=O
Total Metal Impurities
0.5% max.

Description

Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text