Glassy carbon spherical powder, 10-20 micron, type 2, Thermo Scientific Chemicals
Glassy carbon spherical powder, 10-20 micron, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon spherical powder, 10-20 micron, type 2, Thermo Scientific Chemicals

Used as a catalyst support for bimetallic Rh-ReOx. It is also used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 043490.18
also known as 043490-18
Price (USD)/ Each
196.65
Online exclusive
218.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
196.65
Online exclusive
218.00 
Save 21.35 (10%)
Add to cart
Glassy carbon spherical powder, 10-20 micron, type 2, Thermo Scientific Chemicals
Catalog number043490.18
Price (USD)/ Each
196.65
Online exclusive
218.00 
Save 21.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGlassy carbon spherical powder
Recommended StorageAmbient temperatures
EINECS Number231-153-3
TSCAYes
PackagingGlass bottle
View more
Glassy carbon is used as a catalyst support for bimetallic Rh-ReOx. It is also used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only