5-Methylcytosine, 97%, Thermo Scientific Chemicals
5-Methylcytosine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methylcytosine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Catalog number 043492.ME
also known as 043492-ME
Price (USD)/ Each
393.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
393.00
Add to cart
5-Methylcytosine, 97%, Thermo Scientific Chemicals
Catalog number043492.ME
Price (USD)/ Each
393.00
-
Add to cart
Chemical Identifiers
CAS554-01-8
IUPAC Name6-amino-5-methyl-1,2-dihydropyrimidin-2-one
Molecular FormulaC5H7N3O
InChI KeyLRSASMSXMSNRBT-UHFFFAOYSA-N
SMILESCC1=C(N)NC(=O)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (unspecified)> 96.0%
FormPowder
5-Methylcytosine is used as a marker to protect DNA from being cut by native methylation-sensitive restriction enzymes. It is also employed in the production of 4-amino-5-hydroxy-5-methyl-2-oxo-2,5-dihydro-imidazole-1-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only