Thermo Scientific Chemicals

Ammonium tetrathiomolybdate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 043493.06
5 g, Each
Thermo Scientific Chemicals

Ammonium tetrathiomolybdate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 043493.06
5 g, Each
Quantity
Catalog number: 043493.06
also known as 043493-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15060-55-6
IUPAC Name
diammonium disulfanylidenemolybdenum disulfanide
Molecular Formula
H10MoN2S4
InChI Key
ZKKLPDLKUGTPME-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
Appearance (Color)
Black to brown
Assay (special)
?2.5 % of Theoretical value (Mo)
Total Metal Impurities
≤0.05% (500 ppm)
Form
Iridescent crystals

Description

Ammonium tetrathiomolybdate is used as an important reagent in synthetic chemistry and as a building block in bioinorganic chemistry. It is used in the treatment of copper toxicosis in animals and in Wilson's disease. It acts as a chelator in vivo to reduce copper levels. It interferes with angiogenesis and reduces tumor growth. Further, it is used in the synthesis of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands. It is involved in the ring opening of aziridines leading to sulfides and disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text